About this item
The NanoBRET Nano-Glo Detection System provides the NanoBRET Nano-Glo Substrate used by NanoLuc Luciferase to generate the donor signal and the HaloTag NanoBRET 618 Ligand for the fluorescent energy acceptor.
- Dedicated Detection for NanoBRET™ Protein Interaction Assays
- Measure protein interactions in live cells with full-length proteins
- Study both on and off protein interactions with a reversible assay
- Increased signal and lower background compared to conventional BRET assays
The NanoBRET Nano-Glo Detection System provides the NanoBRET Nano-Glo Substrate used by NanoLuc Luciferase to generate the donor signal and the HaloTag NanoBRET 618 Ligand for the fluorescent energy acceptor. The HaloTag NanoBRET 618 Ligand is added directly to the cells during plating, and the NanoBRET Nano-Glo Substrate is added to the sample just prior to measuring donor and acceptor emission. For more details on using the NanoBRET assay for protein:protein interaction studies visit the NanoBRET Technology for Protein Interactions page.
NanoBRET Nano-Glo Detection System, 200 Assays is sufficient for 200 assays in 96-well plates or 500 assays in 384-well plates. Detection System, 1000 Assays is sufficient for 1000 assays in 96-well plates or 2500 assays in 384-well plates. Detection System, 10,000 Assays is sufficient for 10,000 assays in 96-well plates or 25,000 assays in 384-well plates.
The HaloTag NanoBRET 618 Ligand is added directly to the cells during plating, and the NanoBRET Nano-Glo Substrate is added to the sample prior to measuring donor and acceptor emission.
Specifications
- Storage Conditions:Store at –30°C to –10°C and away from light.
- Storage Temperature:-30°C to -10°C