Order Entry
Puerto Rico
ContactUsLinkComponent
 
ISOQUANT® Isoaspartate Detection Kit, Promega
Catalog # PAMA1010
CAS Number:  
undefined
ISOQUANT® Isoaspartate Detection Kit, Promega
Catalog # PAMA1010
Supplier Number:  MA1010
CAS Number:  

Specifications

  • Description:
    ISOQUANT Isoaspartate Detection Kit, 100 assays
  • Size:
    100 assays
  • Storage Conditions:
    Store at –20°C
  • Storage Temperature:
    -30°C to -10°C
  • Cat. No.:
    PAMA1010
  • Supplier no.:
    MA1010

Specifications

About this item

ISOQUANT Isoaspartate Detection Kit is intended for quantitative detection of isoaspartic acid residues in proteins and peptides.

  • Quantitative Detection of Isoaspartic Acid Residues in Proteins and Peptides
  • Monitor Protein Deamidation during Storage and Production
  • Environmental and Industrial Uses

The ISOQUANT Isoaspartate Detection Kit is intended for quantitative detection of isoaspartic acid residues in proteins and peptides, which can result from the gradual, nonenzymatic deamidation of asparagine or rearrangement of aspartic acid residues during storage or handling. Because the kit does not depend on the monitoring of charge differences for detection, charge heterogeneity does not interfere with the assay. The ISOQUANT Kit can be used on peptides or proteins such as monoclonal antibodies.

The ISOQUANT Kit can be used on peptides or proteins such as monoclonal antibodies.