Order Entry
Puerto Rico
ContactUsLinkComponent
Flexi Rabbit Reticulocyte Lysate System, 30 reactions, Promega
Flexi Rabbit Reticulocyte Lysate System, 30 reactions, Promega
Catalog # PAL4540
CAS Number:  
undefined
Flexi Rabbit Reticulocyte Lysate System, 30 reactions, Promega
Catalog # PAL4540
Supplier Number:  L4540
CAS Number:  

Specifications

  • Description:
    Flexi Rabbit Reticulocyte Lysate System, 30 reactions
  • Size:
    30 reactions
  • Storage Conditions:
    Store items at –70° except for Luciferase Assay Wells, which can be stored at room temperature. Do not freeze–thaw the lysate more than two times.
  • Storage Temperature:
    LESS THAN -65°C
  • Cat. No.:
    PAL4540
  • No. of reactions:
    30
  • Supplier no.:
    L4540

Specifications

About this item

Flexi Rabbit Reticulocyte Lysate System allows optimization of translation reactions for a range of parameters, including Mg(2+) and K(+) and DTT concentration.

  • Optimized Translation for Greater Protein Production
  • Increase protein production as much as fourfold over standard lysate reactions
  • For easy optimization the endogenous magnesium concentration is provided for each lot of Flexi(R) Lysate
  • The Flexi(R) System contains three Amino Acid Mixtures, providing a choice of radioisotopes

The Flexi Rabbit Reticulocyte Lysate System allows translation reactions to be optimized for a wide range of parameters, including Mg(2+) and K(+) concentrations and the choice of adding DTT. To help optimize Mg(2+) for a specific message, the endogenous Mg(2+) concentration of each lysate batch is stated in the product information included with this product. The Flexi System also offers the choice of three amino acid mixtures and includes a control RNA encoding the firefly luciferase gene.

The Flexi System also offers the choice of three amino acid mixtures and includes a control RNA encoding the firefly luciferase gene.