Order Entry
Puerto Rico
ContactUsLinkComponent
Glo Lysis Buffer, 1X, Promega
Glo Lysis Buffer, 1X, Promega
Catalog # PAE2661
CAS Number:  
undefined
Glo Lysis Buffer, 1X, Promega
Catalog # PAE2661
Supplier Number:  E2661
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Glo Lysis Buffer (GLB), 1X
  • Size:
    100 mL
  • Storage Conditions:
    Store Glo Lysis Buffer at 4°C. For long–term storage, Glo Lysis Buffer can be frozen at –20°C or –70°C.
  • Cat. No.:
    PAE2661
  • Supplier no.:
    E2661

Specifications

About this item

Glo Lysis Buffer (GLB), 1X, is a proprietary formulation developed to promote rapid lysis of cultured mammalian cells within five minutes without scraping or performing freeze-thaw cycles. 

  • No Need for Cell Scraping or Freeze-Thaw Cycles
  • Cell Lysis within Five Minutes
  • Firefly Luciferase Enzyme in Glo Lysis Buffer Is Stable at Room Temperature for at Least 48 Hours
  • Versatile Buffer Works with Multiple Reagents
  • For Promoting Rapid Lysis of Cultured Mammalian Cells

It is fully compatible with Bright-Glo™, Steady-Glo®, ONE-Glo™ and Renilla-Glo® Luciferase Assay Reagents and the Luciferase Assay Reagent for analysis of firefly luciferase expression.

The half life of these reagents remains the same with or without use of GLB: over five hours for Steady-Glo Reagent and over 24 minutes for Bright-Glo Reagent.