Order Entry
Puerto Rico
ContactUsLinkComponent
Wizard® SV 96 PCR Clean-Up System, Promega
Wizard® SV 96 PCR Clean-Up System, Promega
Catalog # PAA9341
CAS Number:  
undefined
Wizard® SV 96 PCR Clean-Up System, Promega
Catalog # PAA9341
Supplier Number:  A9341
CAS Number:  

Specifications

  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    Wizard SV 96 PCR Clean-Up System, 4 x 96 preps
  • Size:
    4 x 96 preps
  • Storage Conditions:
    15°C to 30°C
  • Description:
    Wizard SV 96 PCR Clean-Up System
  • Cat. No.:
    PAA9341
  • Supplier no.:
    A9341

Specifications

About this item

The Wizard SV 96 PCR Clean-Up System is designed for high-throughput purification of 100 bp to 10 kb PCR products from excess nucleotides, primers, and primer dimers.

  • Optimized Methods Deliver Purified PCR Products Suitable for Demanding Applications Such as Microarray Analysis
  • Average Recovery for 100 to 500 bp Fragments of More than 90%
  • Automated Fluorescent Sequencing Phred Scores of More than 600

This membrane-based system allows recovery of over 90% in as little as 20 minutes

The system uses 96-well filtration without needing to disassemble the manifold. Filtrate waste goes directly to a vacuum trap, which eliminates the need to dispose of collected waste within the manifold assembly.

The purified DNA can be used for automated fluorescent sequencing, cloning, labeling, restriction digestion, or microarray analysis without further manipulation.