Order Entry
Puerto Rico
ContactUsLinkComponent
 
Biotinylated Oligo(dT) Probe (50 pmol/ul), 35 µl, Promega
Catalog # PAZ5261
CAS Number:  
undefined
Biotinylated Oligo(dT) Probe (50 pmol/ul), 35 µl, Promega
Catalog # PAZ5261
Supplier Number:  Z5261
CAS Number:  

Specifications

  • Description:
    Biotinylated Oligo(dT) Probe (50pmol/ul), 35microl
  • Size:
    35 μl
  • Storage Conditions:
    Store at 4 °C. Do not freeze the MagneSphere Paramagnetic Particles.
  • Cat. No.:
    PAZ5261
  • Supplier no.:
    Z5261

Specifications

About this item

The PolyATtract System 1000 isolates messenger RNA directly from crude cell or tissue lysates, eliminating the need for total RNA isolation.

  • Isolates mRNA Directly from Crude Cell or Tissue Lysates
  • mRNA Purification is complete 45 minutes or less
  • Works with tissue amounts from 5mg-2g per isolation
  • No lengthy ethanol precipitation steps, phenol:chloroform extractions, and overnight ultracentrifugation
  • Magnetic separation stand (Cat.# Z5410) accommodates 1.5ml, 2ml, 15ml and 50ml tube sizes

The PolyATtract System 1000 isolates messenger RNA directly from crude cell or tissue lysates, eliminating the need for total RNA isolations. This system uses the MagneSphere technology for the purification of poly(A)+ RNA, eliminating the need for oligo(dT) cellulose columns. The increased yield of mRNA using this system allows the detection of low-copy-number mRNAs in relatively small amounts of material using Northern blot analysis. The isolated mRNA is suitable for all molecular biology applications, including in vitro translation, cDNA synthesis, PCR analysis, ribonuclease (RNase) protection assays, primer extension and Northern blots. The MagneSphere Technology Magnetic Separation Stands can be used in conjunction with any of the PolyATtract Systems and are ideal for applications requiring multiple paramagnetic isolations of biomolecules.