Order Entry
Puerto Rico
ContactUsLinkComponent
 
Flammable
Health
Irritant
BCIP/NBT Color Development Substrate
Catalog # PAS3771
CAS Number:  
undefined
Flammable
Health
Irritant
BCIP/NBT Color Development Substrate
Catalog # PAS3771
Supplier Number:  S3771
CAS Number:  

Specifications

  • Size:
    1.25/2.5 ml
  • Cat. No.:
    PAS3771
  • Supplier no.:
    S3771

Specifications

About this item

 :
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Refrigerator