ContactUsLinkComponent
TNT SP6 Coupled Wheat Germ Extract System, 40 Reactions, Promega
undefined
TNT SP6 Coupled Wheat Germ Extract System, 40 Reactions, Promega
  • Storage Conditions:
    Store the polymerase at –20°C. Store the Luciferase Assay Wells at room temperature. Store the other components at –70°C. Avoid multiple freeze–thaw cycles.
  • Storage Temperature:
    LESS THAN -65°C
Frequently Bought Together