Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Wheat Germ Extract, Promega

Wheat Germ Extract, Promega

Wheat Germ Extract, 5 x 200 μl
Catalog # PAL4380
Supplier:   Promega Corporation
Supplier #:  L4380
Wheat Germ Extract, Promega
Catalog # PAL4380
CAS Number:  
Wheat Germ Extract, Promega
Wheat Germ Extract, Promega

Wheat Germ Extract, 5 x 200 μl

CAS Number:  
Price
$322.51
Log in for availability
Each (1ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Wheat Germ Extract contains the cellular components necessary for protein synthesis (tRNA, ribosomes, initiation, elongation and termination factors) and is used for in vitro translation..

  • Components for Protein Synthesis
  • Contains tRNA, ribosomes, initiation, elongation and termination factors
  • Extract is cleared of endogenous amino acids, mRNA and plant pigments, minimizing background translation

Wheat Germ Extract contains the cellular components necessary for protein synthesis (tRNA, ribosomes, initiation, elongation and termination factors). Wheat Germ Extract is prepared by grinding wheat germ in an extraction buffer followed by centrifugation to remove cell debris. The supernatant is subjected to chromatography that separates endogenous amino acids and plant pigments from the extract. The extract also is treated with micrococcal nuclease to destroy endogenous mRNA and thus reduce background translation to a minimum.

The extract is also treated with micrococcal nuclease to destroy endogenous mRNA and thus reduce background translation to a minimum.

The supernatant is subjected to chromatography that separates endogenous amino acids and plant pigments from the extract.

Documents

Specifications

Specifications

  • Description:
    Wheat Germ Extract
  • Size:
    5 x 200 μl
  • Storage Conditions:
    Store at –70°C or below. Avoid freeze–thaw cycles.
  • Cat. No.:
    PAL4380
  • Storage Temperature:
    LESS THAN -65°C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.