About this item
The TNT T7 Insect Cell Extract Protein Expression System is a convenient, quick, single-tube, coupled transcription/translation system for cell-free protein expression.
- Single-Tube, Coupled Transcription/Translation for Cell-Free Protein Expression using Insect Cell Extract
- Quick, single-tube, coupled transcription/translation system for the cell-free expression of proteins from DNA templates
- Extract derived from Spodoptera frugiperda Sf21 cell line
- Proteins expressed from genes cloned downstream of the T7 promoter; companion vectors available
The TNT T7 Insect Cell Extract Protein Expression System is a convenient, quick, single-tube, coupled transcription/translation system for the cell-free expression of proteins. Protein synthesis reactions are initiated by the addition of a DNA template, eliminating the need for the time-consuming process of in vitro RNA synthesis. The extract is made from the commonly used Spodoptera frugiperda Sf21 cell line. All components necessary for the transcription/translation are present in the TNT T7 ICE Master Mix. To initiate protein synthesis, the only component that must be added is the DNA template. Reactions are incubated at 28-30°C and are complete within 4 hours. Proteins are expressed from genes cloned downstream of the T7 promoter. Companion vectors have been designed to achieve optimal yield with this system. They contain untranslated region (UTR) sequences at the 5' and 3' ends of the gene coding region to enhance translation efficiency. Using the TNT T7 Insect Cell Extract Protein Expression System and these vectors, 75ug/ml of functional protein can be produced.
Proteins are expressed from genes cloned downstream of the T7 promoter. Companion vectors have been designed to achieve optimal yield with this system (pF25A and pF25K). They contain untranslated region (UTR) sequences at the 5´ and 3´ ends of the gene coding region to enhance translation efficiency. With the TnT T7 Insect Cell Extract Protein Expression System and these vectors, 75 μg/mL of functional protein can be produced.
The extract is made from the commonly used Spodoptera frugiperda Sf21 cell line. All components necessary for the transcription/translation are present in the TnT T7 ICE Master Mix. To initiate protein synthesis, the only component that must be added is the DNA template. Reactions are incubated at 28 to 30°C and are complete within four hours.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Temperature:LESS THAN -65°C
- Storage Conditions:Store at –70°C