ContactUsLinkComponent
CellTiter-Glo One Solution Assay, Promega
CellTiter-Glo One Solution Assay, Promega
Certificates
undefined
CellTiter-Glo One Solution Assay, Promega
Certificates
Specifications
  • Storage Conditions:
    Store the CellTiter–Glo One Solution Assay below –10°C. CellTiter–Glo One Solution Assay can be stored at 4°C for 48 hours or at 22°C for 10 hours with ~10–12% loss of activity. CellTiter–Glo One Solution Assay can withstand two additional freeze–thaw cycles after the first thaw with approximately 10% loss of activity with each additional freeze–thaw cycle.
  • Storage Temperature:
    -30°C to -10°C