TMB One Solution, 100 ml, Promega
Catalog # PAG7431
Supplier: Promega Corporation
CAS Number:
Specifications
- Description:TMB One Solution
- Size:100ml
- Storage Conditions:Store at 4°C and away from light.
- Storage Temperature:2…10°C
- Cat. No.:PAG7431
- Supplier no.:G7431
Specifications
About this item
TMB One Solution is a chromagen substrate, 3,3',5,5'-tetramethylbenzidine (TMB) provided in a mildly acidic, nonhazardous buffer for HRP detection in ELISA format.
- Substrate for Horseradish Peroxidase Detection in ELISAs
- Single solution provided ready-to-use; just add, incubate, stop and read
- Low background provides greater assay sensitivity
The substrate is provided as a single solution at a ready-to-use working dilution.
It develops a blue reaction product when oxidized by peroxidase and a yellow reaction product in an endpoint multiwell assay after the addition of an acid solution provided by the end user.
This product is not recommended for Western analysis or immunohistochemistry.