ContactUsLinkComponent
CellTiter 96® Non-Radioactive Cell Proliferation Assay, Promega
CellTiter 96® Non-Radioactive Cell Proliferation Assay, Promega
Certificates
undefined
CellTiter 96® Non-Radioactive Cell Proliferation Assay, Promega
Specifications
  • Storage Conditions:
    Store Dye Solution at –20°C and Solubilization/Stop Solution at room temperature
  • Storage Temperature:
    °COMPONENTS AT DIFFERENT TEMPS
Specifications