About this item
Colistin is a polymyxin antibiotic, it is a mixture of cyclic polypeptides colistin A and B. Colistin is effective against most Gram-negative bacilli and is used as a polypeptide antibiotic.
- Storage temperature: +4 °C, desiccate
Mode of Action: Binds to lipids on the cell cytoplasmic membrane of Gram-negative bacteria and disrupts the cell wall integrity.
Antimicrobial spectrum: Gram-negative bacteria. It is proposed that renal reabsorption of colistin may involve organic cation transporters and peptide transporters and that the process is sensitive to pH.
Colistin sulphate is used to permeabilise bacterial cell membranes and to study mannose-resistant haemagglutination and antibiotic resistance in certain organisms, such as A. baumannii. It has been used to study hephrotoxicity in the rat kidney, and MICs, time-kill kinetics, and postantibiotic effect (PAE) against Pseudomonas aeruginosa.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- CAS No.:1066-17-7
1264-72-8 - Biological Activity:≥15,000 U/mg
- Form:Off-white powder
- MDL No.:MFCD00146495
- Protein Synonyms:Polymyxin E sulfate salt
- Protein/Peptide Name:Colistin sulphate
- Solubility:Soluble in water (50 mg/mL).
- Molecular Formula:C₁₀₄H₂₀₆N₃₂O₄₆S₅