Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Heating Mantle and Control, Quart Can, Glas-Col®

Heating Mantle and Control, Quart Can, Glas-Col®

Heating Mantle, Quart, Can
Catalog # 89259-998
Supplier:   Glas-Col
Supplier #:  100D CH095PL
Heating Mantle and Control, Quart Can, Glas-Col®
Catalog # 89259-998
Supplier:  Glas-Col
Heating Mantle and Control, Quart Can, Glas-Col®
Heating Mantle and Control, Quart Can, Glas-Col®

Heating Mantle, Quart, Can

Price
$1,242.38
Log in for availability
Each
Product Details & Documents

Mantle for quart cans with a basic control.

  • Fabric interior
  • Durable aluminum outer shell
  • Supports weight of quart can

Provides heat and support for a quart can and is used extensively in arson investigation and work done in metal cans with resins and adhesives. Comes complete with compact power control having solid state circuitry, positive on off switches and pilot lights.

Delivery information: 7-10 days after order.

Specifications
  • Capacity:
    Quart
  • Description:
    Heating Mantle
  • For use with:
    Can
  • Cat. No.:
    89259-998
  • Shipping Weight:
    4 lbs.
  • Electrical:
    115V
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.