ContactUsLinkComponent
Lab Stand, Glas-Col
Lab Stand, Glas-Col
Catalog # 58954-025
Supplier:  Glas-Col
Small Business Enterprise
Lab Stand, Glas-Col
Catalog # 58954-025
Supplier:  Glas-Col
Supplier Number:  099DA130150
Small Business Enterprise
Specifications
  • Description:
    Lab Stand, Heavy Duty
  • Cat. No.:
    58954-025
  • For use with:
    Up to 16" dia. Containers
  • Supplier no.:
    099DA130150
Specifications
About this item
Heavy duty lab stand made to last with stainless steel mast and cast iron base.
  • Sturdy construction
  • Accepts up to 16" diameter containers
  • Heavy tip resistant base
This heavy duty lab stand will help to prevent tipping.The balancing weight of the cast iron base and the 3/4" diameter mast make it ideal for use with our stirrers, stir testers and homogenizers.
Delivery: 7-10 days after order.