CRYOBANK™ Bacterial Cultures Freezing and Storing Systems
76263-480
- Description:CRYOBANK™ System with Assorted Colored Beads, 80 Vials
- Cat. No.:76263-480
Cryobank is an easy-to-use and convenient system for freezing and storing bacterial cultures for future retrieval.
- Easy to store: Unique storage boxes fit freezer racks
- Easy to locate: Organism reference grid on lid enables easy indexing and location of cultures
- Versatile: Ready-to-use system for any microbiology applications including: clinical, food and beverage, veterinary, pharmaceutical and biotechnology
- Reliable: Long-term storage and preservation for fastidious organisms
- Convenient: Color-coded system available in four different colors
- Cost-Savings: 64 tube presentation is less expensive than lyophilized commercial cultures
CRYOBANK is based on a cryovial system comprised of chemically treated beads suspended in a cryogenic preserving solution. 25 beads per tube.