Order Entry
Puerto Rico
ContactUsLinkComponent
Lecithin from egg yolk ≥98%
Lecithin from egg yolk ≥98%
Catalog # 100290-566
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Lecithin from egg yolk ≥98%
Catalog # 100290-566
Supplier Number:  370100100MG
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 mg
  • Cat. No.:
    100290-566
  • Supplier no.:
    370100100MG

Specifications

About this item

 :
UN:12352100
ADR:,
CAS Number:93685-90-6
MDL Number:MFCD00131449