Order Entry
Puerto Rico
ContactUsLinkComponent
 
4-Methylumbelliferyl-Β-D-cellobiopyranoside ≥98%, powder
Catalog # 100264-504
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
4-Methylumbelliferyl-Β-D-cellobiopyranoside ≥98%, powder
Catalog # 100264-504
Supplier Number:  M1091100MG
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 mg
  • Cat. No.:
    100264-504
  • Supplier no.:
    M1091100MG

Specifications

About this item

 :
 :500.45 g/mol
UN:12352200
ADR:,
CAS Number:72626-61-0
Formula:C₂₂H₂₈O₁₃
MDL Number:MFCD00063279