Order Entry
Puerto Rico
ContactUsLinkComponent
L-Methionylglycine
L-Methionylglycine
Catalog # 100280-212
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
L-Methionylglycine
Catalog # 100280-212
Supplier Number:  0420891GM
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • CAS No.:
    14486-03-4
  • Size:
    1 g
  • Storage Conditions:
    Store in a cool, dry place
  • Form:
    White fine crystalline powder
  • MDL No.:
    MFCD00021730
  • Protein Synonyms:
    H-Met-Gly-OH
  • Protein/Peptide Name:
    L-Methionylglycine
  • Molecular Weight:
    206.26 g/mol
  • Molecular Formula:
    C₇H₁₄N₂O₃S
  • Cat. No.:
    100280-212
  • Supplier no.:
    0420891GM

Specifications

About this item

  • Melting point: 177 to 178 °C