Exactamix® Empty EVA Bag, Baxter
Supplier: Baxter
Certificates
About this item
EVA (ethylene vinyl acetate) bag, ideal for large-volume sterile solutions.
- Non-reopening clamp attached
- Sterile fluid path
- Non-DEHP
- Non-Latex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.