Cryogenic Vials, Internal or External Threaded, Phenix Research Products
These freestanding heavy-duty sterile cryovials have thick silicone O-ring sets.
- Fast thread design opens or closes in 1¹/₄ turns
- Ridged bottom for one-handed open/close
- Ideal for use in gas phase of LN₂ only
Temperature range : −196 to +121 °C