Closures for Vials, MicroSolv™
PP, White Silicone Rubber/White PTFE, 9-425 mm, Blue, Screw cap
Catalog # 97055-302
Supplier: MicroSolv Technology Corp
Supplier #: 9502S-10M-B
Jump to:
Product Details & Documents
Specifications
- Cap Size:9-425 mm
- Cap type:Screw cap
- Septum:White Silicone Rubber/White PTFE
- Material:PP
- Cap color:Blue