RNase Erase™ Reagent, MP Biomedicals, LLC
Catalog # 75856-412
Supplier: MP Biomedicals
CAS Number:
Specifications
- Description:RNase Erase™
- Application Areas:Nucleic Acid Electrophoresis, Nucleic Acid Sequencing, Nucleic Acid Isolation & Purification
- Storage Temperature:Store at Room temperature (15-30 °C)
- Density:1.000 g/mL at 20 °C(Lit.)
- Product Type:Proteins, Enzymes & Peptides
- Cat. No.:75856-412
- Supplier no.:112440204
Specifications
About this item
Decontamination reagent for RNase.
- Clear, colorless solution
- Ready-to-use
- Effectively removes RNA and RNase contamination
Spray on and rinse off for an RNase-free surface! Completely removes RNase contamination from bench tops, glass and plasticware. RNase Erase is nontoxic and comes packaged with an adjustable spray bottle for for ease of use.