About this item
The stacked arrangement of the cassettes makes easy to load on Tissue-Tek® AutoWrite® Cassette Printer
- Chemically resistant
- Offer textured writing surfaces for clear printing
- Variety of colors choices available
The cassettes are bound by adhesive tape that can easily be removed after the cassettes are inserted into the cassette magazine.