Sani-Plex 128 Detergent, Quiplabs
10121-284
- Description:Sani-Plex 128
- Size:1 gallon
- Cat. No.:10121-284
- Supplier no.:SP128M-CASE
Sani-Plex 128 is a multipurpose, neutral pH, germicidal detergent that disinfects and cleans in one labor-saving step.
It is effective in hard water in the presence of a moderate amount of soil (5% organic serum) as calculated by the AOAC Use-dilution Test.