ContactUsLinkComponent
Masterflex® L/S 24, Pump Tubing, C-Flex®, Cole Parmer
Masterflex® L/S 24, Pump Tubing, C-Flex®, Cole Parmer
  103012-108
Masterflex® L/S 24, Pump Tubing, C-Flex®, Cole Parmer
  103012-108
 :  EW-06427-24
  • Tubing int.Ø:
    6.35 mm (0.250")
  • Description:
    C-Flex® Spooled Pump Tubing
  • Cat. No.:
    103012-108
  • Supplier no.:
    EW-06427-24
It is applicable for general-purpose lab requiring high performance and long life.
  • Biocompatible
  • Optically inspected and factory tested
  • Opaque, white
  • Low gas permeability
Temperature range: –45 to 135 °C