ContactUsLinkComponent
Meiji Techno MA285 Microscope Stage Micrometer, COLE PARMER
Meiji Techno MA285 Microscope Stage Micrometer, COLE PARMER
Catalog # 103163-578
Meiji Techno MA285 Microscope Stage Micrometer, COLE PARMER
Catalog # 103163-578
Supplier Number:  EW-48404-80
Specifications
  • Description:
    Microscope Stage Micrometer, 1 mm Scale in 0.01 mm Divisions
  • Cat. No.:
    103163-578
  • Supplier no.:
    EW-48404-80
Specifications
About this item
Use these micrometers to make quick and accurate measurements of your specimen.