Order Entry
Puerto Rico
ContactUsLinkComponent
Ethanol denatured absolute (200 Proof)
Ethanol denatured absolute (200 Proof)
Catalog # 71003-544
Supplier:  Decon Labs
CAS Number:  
Ethanol denatured absolute (200 Proof)
Catalog # 71003-544
Supplier:  Decon Labs
Supplier Number:  29043A
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 gal.
  • Cat. No.:
    71003-544
  • Supplier no.:
    29043A

Specifications

About this item

 :
 :46.07 g/mol
UN:1170
ADR:3,II
CAS Number:64-17-5
Formula:H₃CCH₂OH
MDL Number:MFCD00003568