BT Lab Systems Tube Roller/Mixer, G-Biosciences
Supplier: G-Biosciences
Certificates
About this item
This tube roller provides two operation modes – rolling and swinging to improve mixing efficiency and safety. It is applied in preventing blood clotting, immunoprecipitation, and other similar applications.
- Compactly designed, simple operation
- LED display time and speed
- Rolling and swinging motion
- Continuous working mode
- Stable performance motor, operation speed adjustable
- Low noise, long life
- Compatible with different sizes of tubes
- 6 rollers
The tube roller is widely used in mixing blood sample, sticky substance and Solid-liquid suspension, etc. Choice of Mixer, Tube Roller with or without Swing.
Specifications
- Catalog No:
- 10856-420
- 10856-418
- Shaking motion:
- 24 mm swing motion
- 24 mm swing motion
- Display:
- LED
- LED