BT Lab Systems Micro-Plate Shaker, G-Biosciences
10856-394
- Orbit:3 mm
- Description:Micro-Plate Shaker
- Timer:99 h, 59 min
- Cat. No.:10856-394
- Supplier no.:BT908
Micro-Plate Shaker, Ideal for ELISA plates (96 and 384 wells) and cell culture plates
- Speed and time under microprocessor control
- Safe and stable with minimal noise
- DC brushless motor, long life, free of maintenance. Can provide gentle and powerful mixing
- Timing function, alarm notification after program completion
Micro-Plate Shaker has LED digital display with simple and easy operation panel that can accurately display the time and the speed of shaking parameters.
Can accommodate 4 standard micro plates.