BT Lab Systems Rocking Shaker, G-Biosciences
10856-392
Irregular Voltage
- Description:Rocking Shaker
- Max. Load:5 kg
- Weight capacity:5 kg
- Platform Size:280×228 mm
- Tilt Angle:10º
- Dimensions:284×264×100 mm
- Material:Stainless steel
- Operating Temperature:4...45 °C
- Timer:1 min. to 99 h, 59 min
- Cat. No.:10856-392
- Electrical:100/240 V, 1.5 A
- Supplier no.:BT907
Rocking Shaker is compactly designed, takes up very little space. It is also used for cell culture and cell membrane transfer.
- Platform made of stainless steel and natural rubber, which is alkali resistant, non deforming, easy-clean and spill-prevention
- Speed real-time display and diminishing time display
- The bearing is articulated rotating, secure and no wear, suitable for prolonged use
- Brushless DC motor, maintenance-free
Rocking Shaker is used in the immobilization of running gels, shaking for dyeing or fading Coomassie blue, immobilization, dyeing and developing of silver-staining, developing and fixation of X-ray film in autoradiography experiments, and further processing of cellulose membrane after gel shift such as molecular hybridization or staining with antigen-antibody.
A benchtop shaker suitable for ambient and incubator incubations.