BT Lab Systems Single Channel Variable Pipettes, G-Biosciences
Supplier: G-Biosciences
Certificates
About this item
Single channel variable volume pipette with choice of volume ranges.
- Ergonomic design ensuring light weight and swift plunger movement
- Calibration as per the standard of EN ISO 8655 for ensured accuracy and precision
- Calibration report enclosed with every pipette
- Click - Stop digital system for volume setting
- Color coded for easy identification
Single channel variable volume pipette with choice of volume ranges. P10: 0.5 to 10 µl; P100: 10 to 100 µl; P1000: 100 to 1000 µl; P2: 0.1 to 2.5 µl; P20: 2 to 20 µl; P200: 20 to 200 µl; or P5000: 1000 to 5000 µl.