Order Entry
Puerto Rico
ContactUsLinkComponent
BT Lab Systems Cell Freezing System, G-Biosciences
BT Lab Systems Cell Freezing System, G-Biosciences
Catalog # 10858-518
Supplier:  G-Biosciences
BT Lab Systems Cell Freezing System, G-Biosciences
Catalog # 10858-518
Supplier:  G-Biosciences
Supplier Number:  BT1401

Specifications

  • Description:
    Cell Freezing System, 2 ml cell suspension/tube maximum volume
  • Cat. No.:
    10858-518
  • Supplier no.:
    BT1401

Specifications

About this item

The CellHome Cell Freezing System, in combination with a -80°C freezer or dry ice locker, will provide the freezing rate of -1°C per minute that is ideal for cryo-preservation of most cultured cell lines.

  • '–1°C per minute in a -80°C environment

The CellHome Cell Freezing System's design uses a combination of insulation foam, radial symmetry, and a heat transfer core to regulate heat loss rather than using a large thermal mass (alcohol based freezer).

As a result, freezing profiles are extremely consistent from one run to the next. Due to the low thermal mass, the system will not cause a rise in local freezer temperature and will protect nearby samples already stored in the freezer. The low thermal mass also means the system will rapidly return commercially available cryogenic storage vials to room temperature.