BT Lab Systems Circular Dry Bath/Heat Block, G-Biosciences
10858-486
- Temperature Range:5°C above ambient to 100°C
- Description:Dry Bath, Circular
- Heating Time:<10 min. from 20 to 100°C
- Cat. No.:10858-486
- Supplier no.:BT1107
A highly economical dry bath incubator.
- Fast and accurate heating
- Precise temperature control
- Built-in temperature deviation calibration function, automatic fault detection, and beep alarm function
- Built-in over-temperature protection device for safety and reliability
- Compact design for small footprint
- Transparent cover
- Several optional circular heating blocks are available
A dry bath incubator that offers excellent temperature accuracy.
: Accessory Heat blocks are not supplied with the equipment and must be ordered separately.
The following blocks are available (See Related Accessories): 24 x 0.5ml and 30 x 1.5ml tubes; 96 x 0.2ml tubes; 59 x 0.5ml tubes; 39 x 1.5ml tubes; 39 x 2.0ml tubes; and 18 x 5.0ml tubes.
The following blocks are available (See Related Accessories): 24 x 0.5ml and 30 x 1.5ml tubes; 96 x 0.2ml tubes; 59 x 0.5ml tubes; 39 x 1.5ml tubes; 39 x 2.0ml tubes; and 18 x 5.0ml tubes.
: No accessory items are supplied with this equipment and must be ordered separately.