Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mini Rotator, Glas-Col
  33725-042
 :  Glas-Col
Small Business Enterprise
undefined
Mini Rotator, Glas-Col
  33725-042
 :  Glas-Col
 :  099AMR1512
Small Business Enterprise

 

  • Shaking motion:
    End over end mixing from a gentle slosh to a rapid agitation
  • Shipping Weight:
    6 lbs.
  • Speed:
    2-80 rpm
  • Cat. No.:
    33725-042
  • Electrical:
    120 V
  • For use with:
    Up to 30 Micro Tubes, 5-9 mm and 8-14 mm
  • Supplier no.:
    099AMR1512

 

 

End over end rotation mixing from a gentle slosh to a rapid agitation for micro centrifuge tubes

  • Ideal smaller size for use on lab bench
  • Interchangeable heads
  • No balancing of load required

Offers optimum mixing from a gentle slosh to a rapid agitation in a compact package making it ideal for use in area where space is a premium.

 : Mini Rotator with aluminum disk head and spring clips for 5-10 mm and 12-14 mm tubes.
 : 7-10 days after order.