Order Entry
Puerto Rico
ContactUsLinkComponent
Holder for Rugged Rotator, 10 to 30 mm Test Tubes, Glas-Col
Holder for Rugged Rotator, 10 to 30 mm Test Tubes, Glas-Col
Catalog # 62404-012
Supplier:  Glas-Col
Small Business Enterprise
undefined
Holder for Rugged Rotator, 10 to 30 mm Test Tubes, Glas-Col
Catalog # 62404-012
Supplier:  Glas-Col
Supplier Number:  099ARD65
Small Business Enterprise

Specifications

  • Capacity:
    Test Tubes, 10 - 30 mm
  • Description:
    Tube Holder Rack, Test Tube
  • Cat. No.:
    62404-012
  • Weight:
    2.7 lbs.
  • Supplier no.:
    099ARD65

Specifications

About this item

Used with the Rugged Rotator for blood agitation, isotopic competitive protein binding assays and similar procedures.

  • Tubes can be inserted radially
  • Equipped with adjustable spring clips
  • Clips designed for side rather than top insertion of tubes

Furnished with 31 spring clips for 10 to 18 mm diameter tubes and 16 spring clips for 20 to 30 mm diameter tubes.

Ordering information: Holder with two sizes of spring clips.
Delivery: 7 to 10 days after order.