Holder for Rugged Rotator, Test Tube, Glas-Col
62404-014
: Glas-Col
Small Business Enterprise
- Capacity:Tissue Culture Tube up to Ø16 mm
- Description:Tube Holder Rack for Test Tube
- Environmentally Preferable:
- Cat. No.:62404-014
- Weight:5 lbs.
- Supplier no.:099ARD10
Frequently used with the Rugged Rotator for embedding tissue for electron microscopy.
- Accepts a variety of glassware
- Tubes easily inserted or removed
- No slip disk design
The spaced disk with springs conveniently holds a combination of test tubes.
: Holder only.
: 7 to 10 days after order.