ContactUsLinkComponent
Accessories for Bench Top Reciprocating Shaker, 75 to 200 mm Centrifuge Tube, Glas-Col
Accessories for Bench Top Reciprocating Shaker, 75 to 200 mm Centrifuge Tube, Glas-Col
  57020-275
 :  Glas-Col
Small Business Enterprise
undefined
Accessories for Bench Top Reciprocating Shaker, 75 to 200 mm Centrifuge Tube, Glas-Col
  57020-275
 :  Glas-Col
 :  099ATR700
Small Business Enterprise
  • Capacity:
    75-200 mm Long
  • Description:
    Tube Holder Rack for Centrifuge Tube
  • Cat. No.:
    57020-275
  • Shipping weight:
    3.5 lbs.
  • Supplier no.:
    099ATR700
This test tube holder lets you shake one or two entire racks of test tubes at once.
  • Holds one or two racks of test tubes
  • Also accomodates bottles, flask or other stoppered containers
Racks are held in place by a restraining cover. This cover automatically releases when retaining nuts are loosened for rack insertion or removal.
 : One holder up to 2 racks, 12 x 5".
 : 7-10 days after order.