ContactUsLinkComponent
POLYVIEW® Plus HRP-DAB (Anti-Rabbit) Kit, Enzo Life Sciences
POLYVIEW® Plus HRP-DAB (Anti-Rabbit) Kit, Enzo Life Sciences
  75841-152
 :  
POLYVIEW® Plus HRP-DAB (Anti-Rabbit) Kit, Enzo Life Sciences
  75841-152
 :  KIT159-0150
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    POLYVIEW® Plus HRP-DAB (Anti-Rabbit) Kit
  • Size:
    150 Tests
  • Environmentally Preferable:
  • Cat. No.:
    75841-152
  • Supplier no.:
    KIT159-0150
Complete staining kit with highest sensitivity, lowest background.
  • Highest sensitivity, complete staining kit for both IHC and ISH
  • POLYVIEW® Plus reagents' biotin free nanopolymer detection circumvents endogenous background while providing specific antigen localization
  • HIGHDEF® DAB (Brown) HRP provides high intensity color development to deliver sharp, crisp staining
  • Compatible with both manual and automated platforms
The POLYVIEW® Plus HRP-DAB (Anti-Rabbit) Kit, is intended for both IHC and ISH staining. The kit is a non-biotin one step detection system suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections. This kit may also be used with blood smears, cytosmears, and cell preparations.
 : Contains IHC/ISH Peroxidase Block, Antibody Blocking Buffer, POLYVIEW® Plus HRP (Anti-Rabbit) Reagent (Ready-To-Use), HIGHDEF® DAB Chromogen, HIGHDEF® DAB Substrate Buffer, HIGHDEF® Hematoxylin, Antigen Retrieval Reagent, pH 9 (10X).