ContactUsLinkComponent
POLYVIEW® Plus AP (Anti-Rabbit) Reagent, Enzo Life Sciences
  75841-148
 :  
POLYVIEW® Plus AP (Anti-Rabbit) Reagent, Enzo Life Sciences
  75841-148
 :  ACC110-0150
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    POLYVIEW® Plus AP (Anti-Rabbit) Reagent
  • Size:
    150 Tests
  • Cat. No.:
    75841-148
  • Supplier no.:
    ACC110-0150
High sensitivity, low background nanopolymer detection reagent for use with HIGHDEF® chromogens in ISH and IHC applications.
  • Ready-to-use reagent
  • Biotin-free nanopolymer detection circumvents endogenous biotin background
  • High intensity color development delivers sharp, crisp staining
  • Compatible with both automated and manual platforms
  • The Antibody Blocker/Diluent reduces background further (sold separately)
  • Customize with HIGHDEF® chromogens of your choice (sold separately)
The POLYVIEW® Plus AP (anti-rabbit) reagent is a ready-to-use, non-biotin, one-step detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the POLYVIEW® Plus AP (anti-rabbit) reagent can be adapted for in situ hybridization detection.