ContactUsLinkComponent
Streptavidin Blocker/Diluent, Enzo Life Sciences
Streptavidin Blocker/Diluent, Enzo Life Sciences
  75841-146
 :  
Streptavidin Blocker/Diluent, Enzo Life Sciences
  75841-146
 :  ACC109-0100
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Streptavidin Blocker/Diluent
  • Size:
    100 mL
  • Cat. No.:
    75841-146
  • Supplier no.:
    ACC109-0100
Ready-to-use blocking buffer and diluent, for use with SAVIEW® Plus detection reagents.
  • Prevent non-specific binding in biotin-conjugated detection systems
  • Intended for qualitative immunohistochemistry
  • May also be used with blood smears, cytosmears, and cell preparations
The Streptavidin Blocker/Diluent can be used to prevent non-specific binding in biotin-conjugated detection systems for IHC and ISH. It can also be used as a diluent for biotinylated antibodies. This product is intended for qualitative immunohistochemistry and in situ hybridization with formalin-fixed paraffin-embedded tissue sections and frozen sections. The blocker/diluent may also be used with blood smears, cytosmears, and cell preparations.