ContactUsLinkComponent
Antibody Blocker/Diluent, Enzo Life Sciences
Antibody Blocker/Diluent, Enzo Life Sciences
Catalog # 75841-144
Supplier:  Enzo Life Sciences
CAS Number:  
Antibody Blocker/Diluent, Enzo Life Sciences
Catalog # 75841-144
Supplier:  Enzo Life Sciences
Supplier Number:  ACC108-0100
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Antibody Blocker/Diluent
  • Size:
    100 mL
  • Cat. No.:
    75841-144
  • Supplier no.:
    ACC108-0100
Specifications
About this item
Ready-to-use antibody blocking buffer and diluent, for use with POLYVIEW® Plus detection reagents.
  • Ready to use
  • Block non-specific staining due to antibodies during immunostaining
  • Intended for immunohistochemistry
The Antibody Blocker/Diluent can be used to block non-specific staining due to antibodies during immunostaining. It can also be used as a diluent for the POLYVIEW® Plus detection reagents or as a primary antibody diluent. This product is intended for immunohistochemistry and in situ hybridization with formalin-fixed parafiin-embedded tissue sections and frozen sections. The blocker/diluent may also be used with blood smears, cytosmears, and cell preparations.