- Description:POLYVIEW® Plus HRP (Anti-Rabbit) Reagent
- Size:150 Tests
- Cat. No.:75841-134
- Supplier no.:ACC103-0150
High sensitivity, low background nanopolymer detection reagent for use with HIGHDEF® chromogens in ISH and IHC applications.
- Ready-to-use reagent
- Biotin-free nanopolymer detection circumvents endogenous biotin background
- High intensity color development delivers sharp, crisp staining
- Compatible with both automated and manual platforms
- The Antibody Blocker/Diluent further reduces background (sold separately)
- Customize with HIGHDEF® chromogens of your choice (sold separately)
The POLYVIEW® Plus HRP (anti-rabbit) reagent is a ready-to-use, biotin-free, one-step detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the POLYVIEW® Plus HRP (anti-rabbit) reagent can be adapted for in situ hybridization detection.