Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trypan Blue
Trypan blue solution 0,5% (w/v) in PBS stain for histology
Trypan blue solution 0,5% (w/v) in PBS stain for histology
Trypan blue solution 0,5% (w/v) in PBS stain for histology
10181-020
:
Avantor
:
Trypan blue solution 0,5% (w/v) in PBS stain for histology
10181-020
:
Avantor
:
Size:
10 ml
Cat. No.:
10181-020
Supplier no.:
PK-CA902-1209
:
:
960.82 g/mol
:
GHS08
:
Ambient
:
72-57-1
:
C₃₄H₂₄N₆Na₄O₁₄S₄
:
MFCD00003969