1510 Disposable N95 Particulate Respirators, Moldex®
Supplier: Moldex/Metric
Certificates
About this item
Heathcare particulate respirators and surgical masks feature a molded nose bridge with soft foam nose cushion to provide the wearer an easy, comfortable, and dependable fit.
- 100% PVC-free
- Dura-Mesh® shell prevents filter media contact by contaminated gloves or hands
- Non-irritative elastic straps are color-coded and securely attached
- Softspun® inner lining is soft to the touch
- Fluid resistant with a bacterial filter efficiency of more than 99.9%
- Available in 5 color-coded sizes
Can be used in surgery, hospitals, and healthcare facilities, nurse/hospice care, correctional facilities, police, fire and rescue teams, EMS/EMT, homeless shelters, occupational health facilities, and school healthcare.
Certifications: Meets CDC guidelines for protection against TB. NIOSH certified to have a filter efficiency of 95% or greater against non-oil based particulates.
Specifications
- Catalog No:
- 89202-646
- 89202-644
- 89202-648
- 89202-640
- 89202-642
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health