Order Entry
Puerto Rico
ContactUsLinkComponent
3-Bladed Stirrers, IKA® Works
3-Bladed Stirrers, IKA® Works
Catalog # 33998-302
Supplier:  IKA Works
undefined
3-Bladed Stirrers, IKA® Works
Catalog # 33998-302
Supplier:  IKA Works
Supplier Number:  477800

Specifications

  • Ø:
    140 mm (head)
  • Description:
    Three-Bladed Propeller Stirrer
  • Cat. No.:
    33998-302
  • Max. speed:
    400 rpm
  • Supplier no.:
    477800
  • Shaft size:
    Ø×L 10×800 mm

Specifications

About this item

For drawing the material to be mixed from the top and the bottom.

  • Made of stainless steel
  • With minimum shearing forces
  • Ideal for stirring liquids at medium to high speeds