Order Entry
Puerto Rico
ContactUsLinkComponent
Corning® Phosphate-Buffered Saline (PBS), 10X Concentrate, pH 7,4, Molecular Biology Grade
Corning® Phosphate-Buffered Saline (PBS), 10X Concentrate, pH 7,4, Molecular Biology Grade
Catalog # 45001-130
Supplier:  Corning
CAS Number:  
undefined
Corning® Phosphate-Buffered Saline (PBS), 10X Concentrate, pH 7,4, Molecular Biology Grade
Catalog # 45001-130
Supplier:  Corning
Supplier Number:  46-013-CM
CAS Number:  

Specifications

  • Description:
    10X Phosphate-Buffered Saline (PBS)
  • Size:
    6 x 1 L
  • Cat. No.:
    45001-130
  • Supplier no.:
    46-013-CM

Specifications

About this item

Phosphate Buffered Saline (PBS) is used to maintain a physiological pH and osmolarity, two key factors in tissue culture and molecular biology research

  • Manufactured in a cGMP environment
  • Formulated with high quality WFI water
  • RNase -, DNase-, and Protease-free
  • PBS without calcium and magnesium
  • pH 7.4 ±0.1
Caution: For research use only.