ContactUsLinkComponent
Difco™ Middlebrook 7H9 Broth, BD Diagnostics
Catalog # 90003-876
Supplier:  BD
CAS Number:  
undefined
Difco™ Middlebrook 7H9 Broth, BD Diagnostics
Catalog # 90003-876
Supplier:  BD
Supplier Number:  271310
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Middlebrook 7H9 Broth
  • Size:
    500 g
  • Cat. No.:
    90003-876
  • Supplier no.:
    271310
Specifications
About this item
Middlebrook 7H9 Broth is recommended for use in qualitative procedures for the isolation and cultivation of Mycobacterium species.
  • Glycerol, albumin, and dextrose support growth
  • Sodium citrate holds certain inorganic cations in solution
  • Ammonium sulfate supplies a nitrogen source
  • Catalase minimizes toxic effects
Middlebrook media consists of many inorganic salts which help in the growth of Mycobacteria. Mycobacteria grow more rapidly in broth media; therefore, primary isolation of all specimens can be done in Middlebrook 7H9 Broth Base.