ContactUsLinkComponent
Difco™ Middlebrook 7H10 Agar, BD
Difco™ Middlebrook 7H10 Agar, BD
  90003-728
 :  BD
 :  
undefined
Difco™ Middlebrook 7H10 Agar, BD
  90003-728
 :  BD
 :  262710
 :  
  • Description:
    Middlebrook 7H10 Agar
  • Size:
    500 g
  • Cat. No.:
    90003-728
  • Supplier no.:
    262710
Middlebrook 7H10 Agar is used in qualitative procedures for the isolation and cultivation of mycobacteria.
  • Inorganic salts provides essential substances
  • Sodium citrate holds certain inorganic cations in solution
  • Glycerol provides plenty of carbon and energy
Supplementation of the agar base is required in order to obtain mycobacterial growth.